siRNA Details
| Gene Symbol: 1700047I17Rik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100116.1 | NP_001093586.1 | 585 | |||
| Gene Symbol: Fam177a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028527.1 | NP_082803.1 | 5 | 585 | ||
| GenBank mRNA | AK006713.1 | - | 418 | |||
| AK045530.1 | - | 645 | ||||
| AK159923.1 | - | 623 | ||||
| BC048158.1 | - | 585 | 5698044 | |||
| BC049669.1 | - | 644 | 6535412 | |||
| Gene Symbol: Srp54c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146826.1 | - | 1712 | |||
| BC005543.1 | - | 1785 | 3585216 | |||

1700047I17Rik2
Genomic Map