siRNA Details
| Gene Symbol: Clcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007711.3 | NP_031737.1 | 10 | 2287 | ||
| NM_173873.1 | NP_776298.1 | 11 | 2463 | |||
| NM_173874.1 | NP_776299.1 | 11 | 2463 | |||
| NM_173876.3 | NP_776301.1 | 10 | 2008 | |||
| GenBank mRNA | AF029347.1 | - | 10 | 1911 | ||
| AK141136.1 | - | 11 | 2401 | |||
| AK147364.1 | - | 9 | 6486 | |||
| AK147548.1 | - | 9 | 6488 | |||
| AK147621.1 | - | 9 | 6488 | |||
| AK149276.1 | - | 11 | 2002 | |||
| AK150688.1 | - | 11 | 2392 | |||
| AK159698.1 | - | 11 | 2437 | |||
| AK161327.1 | - | 11 | 2391 | |||
| AK162639.1 | - | 11 | 2404 | |||
| AK169297.1 | - | 11 | 2384 | |||
| BC057133.1 | - | 10 | 1941 | 6833732 | ||
| BC057855.1 | - | 10 | 2103 | |||
| X78874.1 | - | 10 | 2112 | |||

Clcn3
Genomic Map