siRNA Details
| Gene Symbol: RRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145114.1 | NP_001138586.1 | 2 | 440 | ||
| NM_001284337.1 | NP_001271266.1 | 2 | 440 | |||
| NM_015179.3 | NP_055994.2 | 2 | 440 | |||
| XM_011539555.1 | XP_011537857.1 | 2 | 393 | |||
| XM_011539556.2 | XP_011537858.1 | 2 | 410 | |||
| GenBank mRNA | AB014590.1 | - | 2 | 353 | ||
| AK022496.1 | - | 2 | 411 | |||
| AK296315.1 | - | 2 | 304 | |||
| BC012745.2 | - | 2 | 378 | 4025242 | ||
| DA227012.1 | - | 2 | 410 | |||
| DC417040.1 | - | 2 | 440 | |||

RRP12
Genomic Map