siRNA Details
| Gene Symbol: Chd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081376.1 | NP_001074845.1 | 33 | 5117 | ||
| NM_029216.2 | NP_083492.2 | 32 | 5006 | |||
| GenBank mRNA | AK046510.1 | - | 5 | 549 | ||
| AK129145.1 | - | 9 | 1086 | |||
| AK147563.1 | - | 32 | 5008 | |||
| BC098195.1 | - | 2 | 66 | |||

Chd5
Genomic Map