siRNA Details
| Gene Symbol: Zfp960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005358.2 | NP_001005358.2 | 4 | 424 | ||
| NM_001163919.1 | NP_001157391.1 | 4 | 424 | |||
| GenBank mRNA | AK145357.1 | - | 4 | 412 | ||
| Gene Symbol: Zfp97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011765.5 | NP_035895.3 | 4 | 475 | ||
| GenBank mRNA | CF165138.1 | - | 4 | 475 | ||
| L20450.1 | - | 443 | ||||

Zfp960
Genomic Map