siRNA Details
| Gene Symbol: Vps13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276465.1 | NP_001263394.1 | 39 | 8632 | ||
| NM_001276502.1 | NP_001263431.1 | 38 | 8545 | |||
| GenBank mRNA | AK172952.1 | - | 8 | 1060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13d| Gene Symbol: Vps13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276465.1 | NP_001263394.1 | 39 | 8632 | ||
| NM_001276502.1 | NP_001263431.1 | 38 | 8545 | |||
| GenBank mRNA | AK172952.1 | - | 8 | 1060 | ||