siRNA Details
Gene Symbol: Vps13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276465.1 | NP_001263394.1 | 51 | 10373 | ||
NM_001276502.1 | NP_001263431.1 | 50 | 10289 | |||
GenBank mRNA | AK172952.1 | - | 20 | 2801 | ||
BC037490.1 | - | 6 | 881 |
