siRNA Details
| Gene Symbol: CALCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000728.3 | NP_000719.1 | 5 | 857 | ||
| GenBank mRNA | AK054728.1 | - | 10 | 2079 | ||
| BC092468.1 | - | 5 | 861 | 5313089 | ||
| M28641.1 | - | 1 | 61 | |||
| X02404.1 | - | 3 | 622 | |||

CALCB
Genomic Map