siRNA Details
| Gene Symbol: Sipa1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081028.1 | NP_001074497.1 | 10 | 3084 | ||
| GenBank mRNA | AK154121.1 | - | 10 | 3086 | ||
| AK163313.1 | - | 7 | 1466 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sipa1l3| Gene Symbol: Sipa1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081028.1 | NP_001074497.1 | 10 | 3084 | ||
| GenBank mRNA | AK154121.1 | - | 10 | 3086 | ||
| AK163313.1 | - | 7 | 1466 | |||