siRNA Details
| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177676.6 | NP_808344.3 | 18, 19 | 2450 | ||
| GenBank mRNA | BC137999.1 | - | 14, 15 | 2171 | 8861018 | |
| BC145792.1 | - | 14, 15 | 2173 | 40131095 | ||

4931409K22Rik
Genomic Map