siRNA Details
| Gene Symbol: Ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110218.1 | NP_001103688.1 | 2 | 817 | ||
| NM_176919.4 | NP_795893.2 | 2 | 817 | |||
| GenBank mRNA | AK029461.1 | - | 2 | 379 | ||
| AK040194.1 | - | 2 | 561 | |||
| AK040207.1 | - | 2 | 802 | |||
| AK043121.1 | - | 2 | 818 | |||
| AK051434.1 | - | 2 | 775 | |||
| AK077422.1 | - | 2 | 807 | |||
| AK134804.1 | - | 2 | 797 | |||
| AK150309.1 | - | 2 | 379 | |||
| BC052910.1 | - | 1 | 128 | |||

Ppm1h
Genomic Map