siRNA Details
| Gene Symbol: Sprr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011469.3 | NP_035599.2 | 380 | |||
| GenBank mRNA | AY158987.1 | - | 380 | |||
| AY298855.1 | - | 2 | 454 | |||
| BC132518.1 | - | 366 | 40130795 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr2b| Gene Symbol: Sprr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011469.3 | NP_035599.2 | 380 | |||
| GenBank mRNA | AY158987.1 | - | 380 | |||
| AY298855.1 | - | 2 | 454 | |||
| BC132518.1 | - | 366 | 40130795 | |||