siRNA Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081069.1 | NP_001074538.1 | 11, 12 | 764 | ||
| GenBank mRNA | AF061934.1 | - | 10, 11 | 669 | ||
| AK039314.1 | - | 10, 11 | 1386 | |||
| AK080707.1 | - | 11, 12 | 855 | |||
| AK161551.1 | - | 7, 8 | 823 | |||
| BC049910.1 | - | 9, 10 | 1468 | |||

Rgs11
Genomic Map