siRNA Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133925.2 | NP_598686.2 | 4 | 509 | ||
| GenBank mRNA | AK013047.1 | - | 4 | 479 | ||
| AK019296.1 | - | 4 | 458 | |||
| AK156773.1 | - | 4 | 446 | |||
| AK161670.1 | - | 4 | 474 | |||
| AK167050.1 | - | 4 | 510 | |||
| BC024602.1 | - | 4 | 390 | |||
| BC040811.1 | - | 4 | 454 | 4015830 | ||

Rbm28
Genomic Map