siRNA Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133925.2 | NP_598686.2 | 3, 4 | 484 | ||
| GenBank mRNA | AK013047.1 | - | 3, 4 | 454 | ||
| AK019296.1 | - | 3, 4 | 433 | |||
| AK156773.1 | - | 3, 4 | 421 | |||
| AK161670.1 | - | 3, 4 | 449 | |||
| AK167050.1 | - | 3, 4 | 485 | |||
| BC024602.1 | - | 3, 4 | 365 | |||
| BC040811.1 | - | 3, 4 | 429 | 4015830 | ||

Rbm28
Genomic Map