siRNA Details
| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016780.2 | NP_058060.2 | 3 | 282 | ||
| GenBank mRNA | AF026509.1 | - | 3 | 226 | ||
| AK157958.1 | - | 3 | 228 | |||
| AY541673.1 | - | 3 | 196 | |||
| BC125518.1 | - | 3 | 282 | 40130033 | ||
| BC125520.1 | - | 3 | 282 | 40130035 | ||
| U83167.1 | - | 3 | 193 | |||

Itgb3
Genomic Map