siRNA Details
| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016780.2 | NP_058060.2 | 9 | 1227 | ||
| GenBank mRNA | AF026509.1 | - | 9 | 1171 | ||
| AK156518.1 | - | 5 | 484 | |||
| AK157958.1 | - | 9 | 1173 | |||
| BC125518.1 | - | 9 | 1227 | 40130033 | ||
| BC125520.1 | - | 9 | 1227 | 40130035 | ||

Itgb3
Genomic Map