siRNA Details
| Gene Symbol: Dopey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177208.3 | NP_796182.2 | 5 | 730 | ||
| GenBank mRNA | AK044820.1 | - | 4 | 620 | ||
| AK045873.1 | - | 4 | 617 | |||
| AK052544.1 | - | 4 | 619 | |||
| AK054511.1 | - | 4 | 661 | |||
| AK082462.1 | - | 4 | 653 | |||

Dopey1
Genomic Map