siRNA Details
| Gene Symbol: Dopey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177208.3 | NP_796182.2 | 37 | 7074 | ||
| GenBank mRNA | AK047998.1 | - | 15 | 1580 | ||
| AK169824.1 | - | 10 | 892 | |||
| AK173101.1 | - | 4563 | ||||
| BC035275.1 | - | 7 | 680 | |||
| BC043710.1 | - | 12 | 1295 | |||

Dopey1
Genomic Map