siRNA Details
| Gene Symbol: Kidins220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081378.1 | NP_001074847.1 | 9 | 1048 | ||
| GenBank mRNA | AK083260.1 | - | 3 | 372 | ||
| AK122478.1 | - | 6 | 638 | |||
| BC044772.2 | - | 4 | 404 | |||
| BC046467.1 | - | 4 | 404 | |||
| BC157898.1 | - | 9 | 932 | 9088090 | ||

Kidins220
Genomic Map