siRNA Details
| Gene Symbol: Vmn1r148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030736.2 | NP_109661.1 | 1 | 2327 | ||
| GenBank mRNA | AF324872.1 | - | 1 | 2329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r148| Gene Symbol: Vmn1r148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030736.2 | NP_109661.1 | 1 | 2327 | ||
| GenBank mRNA | AF324872.1 | - | 1 | 2329 | ||