siRNA Details
| Gene Symbol: Mpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025837.2 | NP_080113.1 | 5 | 675 | ||
| GenBank mRNA | AF244360.1 | - | 5 | 657 | ||
| AK003289.1 | - | 3 | 435 | |||
| AK003896.1 | - | 5 | 676 | |||
| AK132777.1 | - | 5 | 676 | |||
| BC009068.1 | - | 5 | 653 | 2650791 | ||
| BC051163.1 | - | 5 | 653 | 6466145 | ||

Mpi
Genomic Map