siRNA Details
| Gene Symbol: Klrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099918.1 | NP_001093388.1 | 5 | 450 | ||
| GenBank mRNA | DQ113419.1 | - | 5 | 450 | ||
| HQ713436.1 | - | 5 | 450 | |||
| JF429832.1 | - | 5 | 463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrb1| Gene Symbol: Klrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099918.1 | NP_001093388.1 | 5 | 450 | ||
| GenBank mRNA | DQ113419.1 | - | 5 | 450 | ||
| HQ713436.1 | - | 5 | 450 | |||
| JF429832.1 | - | 5 | 463 | |||