siRNA Details
| Gene Symbol: Fcgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122603.1 | NP_001116075.1 | 3, 4 | 422 | ||
| GenBank mRNA | AK077773.1 | - | 3, 4 | 423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcgbp| Gene Symbol: Fcgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122603.1 | NP_001116075.1 | 3, 4 | 422 | ||
| GenBank mRNA | AK077773.1 | - | 3, 4 | 423 | ||