siRNA Details
| Gene Symbol: TBC1D12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015188.1 | NP_056003.1 | 5 | 1515 | ||
| XM_006717732.3 | XP_006717795.2 | 5 | 1515 | |||
| XM_006717733.2 | XP_006717796.1 | 6 | 475 | |||
| XM_011539558.2 | XP_011537860.1 | 5 | 526 | |||
| XM_017015997.1 | XP_016871486.1 | 5 | 1515 | |||
| XM_017015998.1 | XP_016871487.1 | 4 | 302 | |||
| XM_017015999.1 | XP_016871488.1 | 4 | 518 | |||
| GenBank mRNA | AB011180.1 | - | 5 | 1331 | ||
| AB449898.1 | - | 5 | 1312 | |||

TBC1D12
Genomic Map