siRNA Details
| Gene Symbol: Srgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081011.2 | NP_001074480.2 | 10, 11 | 2068 | ||
| GenBank mRNA | AK186803.1 | - | 104 | |||
| AY057900.1 | - | 1, 2 | 57 | |||
| BC030457.1 | - | 9, 10 | 1375 | |||
| BC151081.1 | - | 9, 10 | 1686 | 9087994 | ||
| BC151082.1 | - | 9, 10 | 1686 | 9087995 | ||
| BC158055.1 | - | 9, 10 | 1684 | 9088163 | ||
| BC172152.1 | - | 9, 10 | 1686 | 9054822 | ||

Srgap2
Genomic Map