siRNA Details
| Gene Symbol: Stag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009282.3 | NP_033308.2 | 25 | 2828 | ||
| GenBank mRNA | BC062954.1 | - | 25 | 2828 | 30537388 | |
| BC069841.1 | - | 26 | 2756 | |||
| Z75332.1 | - | 25 | 2877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stag1| Gene Symbol: Stag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009282.3 | NP_033308.2 | 25 | 2828 | ||
| GenBank mRNA | BC062954.1 | - | 25 | 2828 | 30537388 | |
| BC069841.1 | - | 26 | 2756 | |||
| Z75332.1 | - | 25 | 2877 | |||