siRNA Details
| Gene Symbol: Lhfpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026571.2 | NP_080847.2 | 1 | 504 | ||
| GenBank mRNA | BC099483.1 | - | 1 | 402 | 30848018 | |
| BC125264.1 | - | 1 | 341 | |||
| BC147842.1 | - | 1 | 302 | 9087559 | ||
| BC147847.1 | - | 1 | 302 | 9087564 | ||

Lhfpl5
Genomic Map