siRNA Details
| Gene Symbol: Khdc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033904.1 | NP_001029076.1 | 2 | 599 | ||
| GenBank mRNA | BC099381.1 | - | 2 | 599 | 30937206 | |
| BC141037.1 | - | 293 | 9055649 | |||
| Gene Symbol: Khdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183322.2 | NP_899145.2 | 2 | 615 | ||
| GenBank mRNA | AK136166.1 | - | 2 | 542 | ||
| AY238603.1 | - | 2 | 378 | |||

Khdc1c
Genomic Map