siRNA Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081045.1 | NP_001074514.1 | 13 | 3426 | ||
| GenBank mRNA | AK006970.1 | - | 5 | 537 | ||
| AK122484.1 | - | 13 | 3432 | |||
| AK161032.1 | - | 2 | 1198 | |||
| BC025052.1 | - | 3 | 307 | |||
| BC043121.1 | - | 13 | 2975 | 6409084 | ||
| BC054752.1 | - | 14 | 3249 | 6834541 | ||
| BC079594.1 | - | 13 | 2974 | 6827379 | ||

Kansl1
Genomic Map