siRNA Details
| Gene Symbol: DNAJC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015190.4 | NP_056005.1 | 3 | 623 | ||
| GenBank mRNA | AF327347.1 | - | 3 | 530 | ||
| AK074420.1 | - | 3 | 559 | |||
| AK094162.1 | - | 3 | 2154 | |||
| BC136507.1 | - | 3 | 550 | 9020496 | ||

DNAJC9
Genomic Map