siRNA Details
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039104.2 | NP_001034193.2 | 12 | 1758 | ||
NM_018752.3 | NP_061222.3 | 12 | 1651 | |||
GenBank mRNA | AF047714.1 | - | 12 | 1520 | ||
AK044507.1 | - | 12 | 1682 | |||
AK148193.1 | - | 10 | 1600 | |||
AK148268.1 | - | 12 | 1505 | |||
AK148275.1 | - | 12 | 1561 | |||
AK149290.1 | - | 12 | 1680 | |||
AK189982.1 | - | 2 | 79 | |||
AK190619.1 | - | 2 | 188 | |||
AK192027.1 | - | 2 | 79 | |||
AK194126.1 | - | 2 | 79 | |||
AK196486.1 | - | 2 | 79 | |||
AK199874.1 | - | 2 | 79 | |||
AK201989.1 | - | 2 | 188 | |||
AK202317.1 | - | 2 | 80 | |||
AK203960.1 | - | 2 | 78 | |||
AK204100.1 | - | 2 | 79 | |||
AK205137.1 | - | 2 | 85 | |||
AK217903.1 | - | 2 | 79 | |||
AY180104.2 | - | 12 | 1758 |
