siRNA Details
| Gene Symbol: Xlr4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081642.1 | NP_001075111.1 | 9 | 1009 | ||
| GenBank mRNA | BC098242.1 | - | 9 | 989 | 1478336 | |
| Gene Symbol: Xlr4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183094.3 | NP_898917.2 | 9 | 1039 | ||
| GenBank mRNA | AA190046.1 | - | 1 | 80 | ||
| AK166871.1 | - | 9 | 1040 | |||

Xlr4a
Genomic Map