siRNA Details
| Gene Symbol: Xlr4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081642.1 | NP_001075111.1 | 3 | 299 | ||
| GenBank mRNA | BC098242.1 | - | 3 | 279 | 1478336 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xlr4a| Gene Symbol: Xlr4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081642.1 | NP_001075111.1 | 3 | 299 | ||
| GenBank mRNA | BC098242.1 | - | 3 | 279 | 1478336 | |