siRNA Details
| Gene Symbol: Gm2002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100596.1 | NP_001094066.1 | 14 | 2297 | ||
| Gene Symbol: Il11ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010550.3 | NP_034680.3 | 14 | 2297 | ||

Gm2002
Genomic Map