siRNA Details
| Gene Symbol: Nek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010892.3 | NP_035022.2 | 2 | 379 | ||
| GenBank mRNA | AK077627.1 | - | 2 | 343 | ||
| AK147072.1 | - | 2 | 355 | |||
| AK164467.1 | - | 2 | 322 | |||
| AK169330.1 | - | 3 | 270 | |||
| BC057576.1 | - | 2 | 319 | 6810914 | ||
| CF738658.1 | - | 2 | 408 | |||

Nek2
Genomic Map