siRNA Details
| Gene Symbol: Gm6880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099305.1 | NP_001092775.1 | 3 | 535 | ||
| Gene Symbol: Gm6890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099306.1 | NP_001092776.1 | 3 | 535 | ||

Gm6880
Genomic Map