siRNA Details
| Gene Symbol: Ankrd44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081433.3 | NP_001074902.2 | 7 | 880 | ||
| GenBank mRNA | AK038343.1 | - | 7 | 881 | ||
| AK157584.1 | - | 7 | 849 | |||
| AK163222.1 | - | 4 | 518 | |||
| BC157918.1 | - | 8 | 698 | 9007346 | ||
| BC157951.1 | - | 7 | 698 | 9007282 | ||
| BC172102.1 | - | 7 | 698 | 9054388 | ||

Ankrd44
Genomic Map