siRNA Details
| Gene Symbol: Ankrd44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081433.3 | NP_001074902.2 | 19 | 2208 | ||
| GenBank mRNA | AK053404.1 | - | 4 | 384 | ||
| BC157918.1 | - | 20 | 2026 | 9007346 | ||
| BC157951.1 | - | 19 | 2026 | 9007282 | ||
| BC172102.1 | - | 19 | 2026 | 9054388 | ||

Ankrd44
Genomic Map