siRNA Details
| Gene Symbol: Scgb1b24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099329.2 | NP_001092799.1 | 2 | 88 | ||
| GenBank mRNA | GU269232.1 | - | 2 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1b24| Gene Symbol: Scgb1b24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099329.2 | NP_001092799.1 | 2 | 88 | ||
| GenBank mRNA | GU269232.1 | - | 2 | 88 | ||