siRNA Details
| Gene Symbol: Erdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133362.2 | NP_579940.1 | 735 | |||
| GenBank mRNA | AJ007909.1 | - | 661 | |||
| AJ539223.1 | - | 673 | ||||
| BC018296.1 | - | 740 | 3499536 | |||
| BC058113.1 | - | 844 | 6820436 | |||
| BC080795.1 | - | 590 | 5006491 | |||
| BC086616.1 | - | 3254 | ||||

Erdr1
Genomic Map