siRNA Details
| Gene Symbol: Erdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133362.2 | NP_579940.1 | 700 | |||
| GenBank mRNA | AK051407.1 | - | 1200 | |||
| BC018296.1 | - | 705 | 3499536 | |||
| BC038133.1 | - | 514 | ||||
| BC058113.1 | - | 809 | 6820436 | |||
| BC080795.1 | - | 555 | 5006491 | |||
| BC082282.1 | - | 652 | ||||
| BC086616.1 | - | 3219 | ||||

Erdr1
Genomic Map