siRNA Details
| Gene Symbol: Cisd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085500.2 | NP_001078969.2 | 3 | 708 | ||
| GenBank mRNA | BC056217.1 | - | 3 | 703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cisd3| Gene Symbol: Cisd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085500.2 | NP_001078969.2 | 3 | 708 | ||
| GenBank mRNA | BC056217.1 | - | 3 | 703 | ||