siRNA Details
| Gene Symbol: Tiam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122998.1 | NP_001116470.1 | 27 | 5070 | ||
| NM_001286757.1 | NP_001273686.1 | 13 | 2113 | |||
| NM_001286758.1 | NP_001273687.1 | 13 | 1477 | |||
| NM_011878.2 | NP_036008.2 | 27 | 5054 | |||
| GenBank mRNA | AB022915.1 | - | 27 | 5055 | ||
| AK129474.1 | - | 27 | 4973 | |||
| AK158489.1 | - | 13 | 1479 | |||
| AK161947.1 | - | 13 | 2114 | |||
| BC067048.1 | - | 27 | 5067 | 30535152 | ||
| BC079600.1 | - | 27 | 4959 | 6853027 | ||

Tiam2
Genomic Map