siRNA Details
| Gene Symbol: KIAA0922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131007.1 | NP_001124479.1 | 20, 21 | 2168 | ||
| NM_015196.3 | NP_056011.3 | 20, 21 | 2165 | |||
| XM_011531780.1 | XP_011530082.1 | 20, 21 | 2218 | |||
| XM_011531781.1 | XP_011530083.1 | 20, 21 | 2215 | |||
| XM_011531783.1 | XP_011530085.1 | 18, 19 | 1966 | |||
| XM_017007924.1 | XP_016863413.1 | 19, 20 | 2140 | |||
| XM_017007925.1 | XP_016863414.1 | 20, 21 | 2168 | |||
| XM_017007926.1 | XP_016863415.1 | 20, 21 | 2165 | |||
| XM_017007927.1 | XP_016863416.1 | 3, 4 | 273 | |||
| GenBank mRNA | AK092270.1 | - | 20, 21 | 2165 | ||
| AL136932.1 | - | 9, 10 | 1125 | |||
| BC044932.1 | - | 13, 14 | 1447 | |||
| BC131505.1 | - | 19, 20 | 1937 | 40082592 | ||

KIAA0922
Genomic Map