siRNA Details
| Gene Symbol: Whsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081102.2 | NP_001074571.2 | 15, 16 | 3028 | ||
| NM_175231.2 | NP_780440.2 | 15, 16 | 3025 | |||
| GenBank mRNA | AK078622.1 | - | 12, 13 | 1664 | ||
| AK129287.1 | - | 12, 13 | 1963 | |||
| BC046473.1 | - | 10, 11 | 1454 | |||
| BC053454.1 | - | 4, 5 | 577 | |||
| BC079668.1 | - | 15, 16 | 3051 | |||

Whsc1
Genomic Map