siRNA Details
| Gene Symbol: Adgrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144045.1 | - | 2 | 433 | ||
| BC132476.1 | - | 2 | 281 | 40130753 | ||
| BC132480.1 | - | 2 | 281 | 40130757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adgrg4| Gene Symbol: Adgrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144045.1 | - | 2 | 433 | ||
| BC132476.1 | - | 2 | 281 | 40130753 | ||
| BC132480.1 | - | 2 | 281 | 40130757 | ||