siRNA Details
| Gene Symbol: KIAA0922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131007.1 | NP_001124479.1 | 33 | 4441 | ||
| NM_015196.3 | NP_056011.3 | 33 | 4438 | |||
| XM_011531780.1 | XP_011530082.1 | 33 | 4491 | |||
| XM_011531781.1 | XP_011530083.1 | 33 | 4488 | |||
| XM_011531783.1 | XP_011530085.1 | 31 | 4239 | |||
| XM_017007924.1 | XP_016863413.1 | 32 | 4413 | |||
| XM_017007925.1 | XP_016863414.1 | 33 | 4438 | |||
| XM_017007926.1 | XP_016863415.1 | 33 | 4435 | |||
| XM_017007927.1 | XP_016863416.1 | 16 | 2546 | |||
| GenBank mRNA | AB023139.1 | - | 11 | 1929 | ||
| AK001454.1 | - | 9 | 1464 | |||
| AK096538.1 | - | 13 | 2258 | |||
| AK128028.1 | - | 2 | 4042 | |||
| AL136932.1 | - | 22 | 3398 | |||
| BC044932.1 | - | 26 | 3720 | |||
| BC131505.1 | - | 32 | 4210 | 40082592 | ||

KIAA0922
Genomic Map