siRNA Details
| Gene Symbol: Atic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026195.3 | NP_080471.2 | 6 | 530 | ||
| GenBank mRNA | AK010449.1 | - | 6 | 499 | ||
| AK010611.2 | - | 6 | 513 | |||
| AK012074.1 | - | 6 | 467 | |||
| AK046353.1 | - | 6 | 513 | |||
| AK051020.1 | - | 508 | ||||
| AK139235.1 | - | 6 | 465 | |||
| AK186275.1 | - | 3 | 219 | |||
| BC039925.1 | - | 6 | 536 | 3155580 | ||

Atic
Genomic Map