siRNA Details
| Gene Symbol: Siglec15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101038.1 | NP_001094508.1 | 6 | 923 | ||
| GenBank mRNA | AY971517.1 | - | 6 | 923 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Siglec15| Gene Symbol: Siglec15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101038.1 | NP_001094508.1 | 6 | 923 | ||
| GenBank mRNA | AY971517.1 | - | 6 | 923 | ||